EP022510070
Eppendorf® IsoRack Starter Set
IsoRack Starter Set for Flex Bucket
About This Item
UNSPSC Code:
41122804
Productos recomendados
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022510070
Categorías relacionadas
General description
IsoRack starter set for Flex bucket, 2 × IsoRack adapters, 2 × IsoRacks with lid, 2 × cool packs (0 °C IsoPack), for 0.5 mL and 1.5/2.0 mL reaction vessels
Legal Information
Eppendorf is a registered trademark of Eppendorf AG