Documentos clave
C4551
Sigma-Aldrich
Vertical mini-gel Comb
10 cm wide, for use with 1.0 mm x 10 wells
About This Item
UNSPSC Code:
41105305
NACRES:
NB.51
product name
Combs for vertical mini-gel, 10 cm wide, size 1.0 mm × 10 wells
size
1.0 mm × 10 wells