Merck
392693
Sigma-Aldrich
Ammonia solution
2.0 M in isopropanol
About This Item
Fórmula lineal:
NH3
Número de CAS:
Peso molecular:
17.03
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55
vapor pressure
20.22 psi ( 55 °C)
6.34 psi ( 20 °C)
form
liquid
concentration
2.0 M in isopropanol
2.0-2.2 M (by NaOH, titration)
bp
50 °C
density
0.773 g/mL at 25 °C
SMILES string
N
InChI
1S/H3N/h1H3
InChI key
QGZKDVFQNNGYKY-UHFFFAOYSA-N
General description
Ammonia solution (ammonia in isopropanol) is a colorless gas with a characteristic strong odor. It is highly soluble in water, alcohol, chloroform ether, ethanol, and methanol. Ammonia solution is commonly used as a base.
Application
Ammonia solution is used for the synthesis of silica particles.
signalword
Danger
Hazard Classifications
Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Central nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
39.2 °F - closed cup
flash_point_c
4 °C - closed cup
Visite la Librería de documentos
Eagleson M.
Concise Encyclopedia Chemistry, 65-66 (1994)
Ledgard J.
The Preparatory Manual of Explosives., 41-41 (2007)
Patrick M Erwin et al.
The ISME journal, 8(3), 575-588 (2013-10-25)
Ascidians are ecologically important components of marine ecosystems yet the ascidian microbiota remains largely unexplored beyond a few model species. We used 16S rRNA gene tag pyrosequencing to provide a comprehensive characterization of microbial symbionts in the tunic of 42
S F Chew et al.
Journal of fish biology, 84(3), 603-638 (2014-01-21)
With the development of air-breathing capabilities, some fishes can emerge from water, make excursions onto land or even burrow into mud during droughts. Air-breathing fishes have modified gill morphology and morphometry and accessory breathing organs, which would tend to reduce
Patricia A Wright et al.
Respiratory physiology & neurobiology, 184(3), 231-240 (2012-08-23)
In this review we pose the following seven questions related to ammonia and fish that represent gaps in our knowledge. 1. How is ammonia excretion linked to sodium uptake in freshwater fish? 2. How much does branchial ammonia excretion in