Z283835
®Pulgas agitadoras magnéticas Spinbar
white, diam. 3 mm, L 3 mm
About This Item
Código UNSPSC:
41103806
NACRES:
NB.72
Productos recomendados
Materiales
PTFE-coated
cylindrical PTFE-coated
Características
ALNICO V coated magnet
envase
pack of 3 ea
fabricante / nombre comercial
Bel-Art F37119-0003
Longitud
3 mm
Diámetro
3 mm
color
white
Categorías relacionadas
Descripción general
- Spinbar® magnetic stirring fleas are designed for use in tubes, vials, and gradient markers.
- Ideal for mixing of small volume samples in environmental testing and life science applications.
- Alnico magnet encapsulated in durable and inert FDA grade PTFE (polytetrafluoroethylene) coating.
- Broad chemical resistance for use in a wide variety of applications.
- Autoclavable.
Very small, efficient stirring bars. Ideal for vials, micro test tubes, centrifuge tube mixing etc.
- PTFE-coated
- ALNICO V coated magnet
Información legal
Spinbar is a registered trademark of SP Scienceware, a division of SP
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)