Documentos clave
Z100609
Sigma-Aldrich
Single-bulb ball-tube
capacity 250 mL, joint: ST/NS 14/20, plain
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
joint
joint: ST/NS14/20
capacity
250 mL
General description
Arranged horizontally outside of Kugelrohr oven, the ball-tubes cool and collect distillate with minimal loss of material.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)