Skip to Content
Merck
All Photos(1)

Key Documents

1087869

USP

Candesartan cilexetil Related Compound F

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Ethoxy-1-[[2′-(2-ethyl-2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylic acid 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H38N6O6
CAS Number:
Molecular Weight:
638.71
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

candesartan

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[n]1(nnc(n1)c2c(cccc2)c3ccc(cc3)C[n]4c5c(nc4OCC)cccc5C(=O)OC(OC(=O)OC6CCCCC6)C)CC

InChI

1S/C35H38N6O6/c1-4-41-38-32(37-39-41)28-15-10-9-14-27(28)25-20-18-24(19-21-25)22-40-31-29(16-11-17-30(31)36-34(40)44-5-2)33(42)45-23(3)46-35(43)47-26-12-7-6-8-13-26/h9-11,14-21,23,26H,4-8,12-13,22H2,1-3H3

InChI key

IPKUBVHJAZIDCZ-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Candesartan cilexetil Related Compound F USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Candesartan Cilexetil
  • Candesartan Cilexetil Tablets
  • Candesartan Cilexetil and Hydrochlorothiazide Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service