Z743619
Laser Tough-Tags™
assorted colors, L × W 0.94 in. × 0.50 in., pkg of 2975
Synonym(s):
solvent resistant labels, temperature resistant labels
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
packaging
pkg of 2975
manufacturer/tradename
Diversified Biotech RNBW-3100
technique(s)
PCR: suitable
L × W
0.94 in. × 0.50 in.
color
assorted colors
Looking for similar products? Visit Product Comparison Guide
General description
Tough-Tags are chemically inert labels that strongly adhere to microtubes, test tubes, storage boxes and other microscope slides. They are temperature resistant from 121°C to –40°C and withstand autoclaving, boiling water baths, freezer temperatures, organic solvents and caustic agents. For best adhesion it is recommended that siliconized surfaces be avoided.Tough-Tags are perfect for accurately identifying important samples stored in tubes, microplates or boxes that will be autoclaved or stored in moderate freezer temperatures.
Legal Information
Tough-Tags is a trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service