SML3405
NVP-LDZ982
≥98% (HPLC)
Synonym(s):
LDZ-982, LDZ 982, LDZ982, Methyl 5-[1-(3-chlorophenyl)-5-(trifluoromethyl)-1H-pyrazol-4-yl]-3-(2,6-dichlorophenyl)isoxazole-4-carboxylate, NITD 982, NITD-982, NITD982
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
color
, Light yellow to orange
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
Biochem/physiol Actions
NVP-LDZ982 (NITD-982) is a dihydroorotate dehydrogenase (DHODH) inhibitor (Kd = 27 nM) with broad-spectrum antiviral activity by targeting host cell de novo pyrimidine biosynthesis pathway. NITD-982 inhibits various viral replications in cultues, including Dengue (DENV EC50 = 5-2.6 nM; A549), hepatitis C virus (HCV EC50 = 7-12.6 nM; Huh-7), HIV (MDR769 EC50 = 0.97 nM; PBMC), Influenza (EC50 = 32 nM/H1N1, 117 nM/B, 217 nM/H1N3; MDCK), respiratory syncytial virus (RSV EC50 = 7-9 nM; Hep-2), and Yellow fever (strain 17D EC50 = 8.2 nM; A549).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Inhibition of dengue virus through suppression of host pyrimidine biosynthesis
Journal of Virology, 85(13), 6548-6556 (2011)
Identification of broad-spectrum antiviral compounds and assessment of the druggability of their target for efficacy against respiratory syncytial virus (RSV)
Proceedings of the National Academy of Sciences of the USA, 108(17), 6739-6744 (2011)
Antimicrobial agents and chemotherapy, 59(4), 2086-2093 (2015-01-28)
Dengue virus (DENV) is the most prevalent mosquito-borne viral pathogen in humans. Currently, there is no clinically approved vaccine or antiviral for DENV. Combination therapy is a common practice in antiviral treatment and a potential approach to search for new
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service