Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 150 g
manufacturer/tradename
NIST®
format
matrix material
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
This Standard Reference Material (SRM) is intended primarily for use in validation of chemical and instrumental methods of analysis used to determine elements in steel alloys. It can be used to validate value assignment of in-house reference materials. A unit of SRM 126c consists of a bottle containing approximately 150 g of chips. For more information, please refer to the COA and SDS.
SRM 126C_cert
SRM 126C_SDS
SRM 126C_cert
SRM 126C_SDS
Other Notes
Certified for the analytes listed below.
Carbon (C), Manganese (Mn), Nickel (Ni), Silicon (Si)
Reference/Informational value is provided for the following analytes.
Cobalt (Co), Chromium (Cr), Copper (Cu), Molybdenum (Mo), Phosphorous (P), Sulfur (S), Vanadium (V)
See certificate for values and more details at nist.gov/SRM.
Carbon (C), Manganese (Mn), Nickel (Ni), Silicon (Si)
Reference/Informational value is provided for the following analytes.
Cobalt (Co), Chromium (Cr), Copper (Cu), Molybdenum (Mo), Phosphorous (P), Sulfur (S), Vanadium (V)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service