Skip to Content
Merck
All Photos(1)

Key Documents

CLS44232XL

Pyrex® Fernbach baffled culture flask

with wide beaded mouth (without stopper)

Synonym(s):

Baffled culture flask, Fernbach flask

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104

material

borosilicate glass (code 7740)
flat bottom flask
wide-mouth
wide-mouth neck

description

Ht: 230mm, Outer Diameter (OD): 200mm, Neck OD: 68mm

sterility

non-sterile

feature

cap: no

packaging

case of 3

manufacturer/tradename

Corning 4423-2XL

capacity

2,800 mL

Looking for similar products? Visit Product Comparison Guide

General description

This 2800mL wide mouth PYREX® Fernbach-style culture flask is designed for culturing organisms requiring a large surface area to volume ratio. It can also be used in serum production. Triple baffled on the bottom outside edges to achieve maximal oxygen transfer to culture medium. Beaded neck rim is approximately 60mm I.D. and accepts cotton plugs or No. 13 rubber stoppers. This flask has a large marking spot.
Note: Do not use this flask on a heat source smaller than the base of the flask. Uneven heating of the flask could cause breakage.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service