Key Documents
BR114730
BRAND® grip stopper
for sample tubes BR114750/BR114760
Synonym(s):
Accessories, Stoppers, Test tubes, Tubes
About This Item
UNSPSC Code:
41100000
NACRES:
NB.11
material
low-density polyethylene
packaging
pack of 1000 ea
manufacturer/tradename
BRAND 114730
Related Categories
General description
The BRAND® Grip Stopper is designed specifically for use with tubes 114750 and 114760, providing a secure and reliable sealing solution. Made from neutral PE-LD (low-density polyethylene), this stopper ensures compatibility with a variety of substances while maintaining a tight seal to prevent contamination. Its ergonomic design allows for easy handling and application, making it ideal for laboratory settings. This grip stopper enhances the functionality of your tubes, ensuring safe and effective storage and transport of liquids and samples.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)