Documentos clave
Z640077
Eppendorf® epT.I.P.S. standard
volume range 0.1-20 μL
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
L:
40 mm
maximum volume:
20 μL
volume range:
0.1-20 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 1000 ea (2 bags of 500 tips)
manufacturer/tradename:
Eppendorf® 0030 000.838
Eppendorf® 022492012
En este momento no podemos mostrarle ni los precios ni la disponibilidad
material
gray
sterility
non-sterile
feature
barrier: no
packaging
pack of 1000 ea (2 bags of 500 tips)
manufacturer/tradename
Eppendorf® 0030 000.838
Eppendorf® 022492012
L
40 mm
maximum volume
20 μL
volume range
0.1-20 μL
General description
Bulk packed in re-sealable bags.
200 μL, 300 μL and 1000 μL tips are color coded, yellow, orange and blue respectively.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)