OH30038555
Ohaus® Reference Electrode
STREF2
About This Item
Código UNSPSC:
39121436
Productos recomendados
envase
pack of 1 ea
fabricante / nombre comercial
Ohaus® 30038555
ORP measuring range
-1000-+1000 mV
Descripción general
All electrodes include a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
Aplicación
pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), temperature measurement
Características y beneficios
Can be used in conjunction with all Starter bench and portable meters
Información legal
Ohaus is a registered trademark of Ohaus Corp.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)