Key Documents
EP022653008
Rotor for Eppendorf® 5424/5424R Centrifuges
FA-45-24-11, aerosol tight, lid, to hold, 24 x 1.5/2.0 mL tubes
About This Item
UNSPSC Code:
41103909
feature
lid
to hold 24 x 1.5/2.0 mL tubes
FA-45-24-11, aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 0022653008 (North America)
parameter
15,000 max. RCF (21,130 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5424/5424R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG