Key Documents
EP0030129342
Eppendorf® Twin tec microbiology PCR plates, 384 wells
384, skirted, colorless plate, conical bottom colorless wells, PCR clean, pkg of 10 ea, colorless
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material
colorless plate
conical bottom colorless wells
feature
PCR clean
skirt
384, skirted
packaging
pkg of 10 ea
manufacturer/tradename
Eppendorf® 0030129342
size
384 wells
color
colorless
Legal Information
Eppendorf is a registered trademark of Eppendorf AG