Key Documents
ADF457282-EP
Adapter for Eppendorf® F-45-72-8 Rotor
holds 1 x .2 mL PCR tube, pack of 6 ea
About This Item
UNSPSC Code:
41103911
feature
holds 1 x .2 mL PCR tube
packaging
pack of 6 ea
manufacturer/tradename
Eppendorf® 022636286 (NA)
technique(s)
PCR: suitable
centrifugation: suitable
compatibility
for use with Rotor F-45-72-8
Categorías relacionadas
Legal Information
Eppendorf is a registered trademark of Eppendorf AG