XXSPTFF03
Pellicon® Single-Pass TFF Cassette Retentate Collection Plate
About This Item
Código UNSPSC:
23151800
eCl@ss:
32031690
Productos recomendados
Materiales
316L stainless steel housing
Nivel de calidad
técnicas
tangential flow filtration (TFF): suitable
Adaptador
3/4 in. sanitary flange drain/vent (TC)
aplicaciones
liquid formulation
Descripción general
The Pellicon Single-Pass TFF Cassette Retentate Collection Plate is required to collect the retentate flow of single-pass TFF installations with Pellicon 3 0.57 m2, Pellicon 3 1.14 m2 or Pellicon 2 0.5 m2 cassettes.
Información legal
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany