Merck
Z117005
Stainless steel 304 syringe needle
gauge 14, L 2 in.
About This Item
UNSPSC Code:
41122002
NACRES:
NB.03
En este momento no podemos mostrarle ni los precios ni la disponibilidad
description
Standard Hub with 304 Stainless Steel Tubing
packaging
pack of 1 ea
manufacturer/tradename
Cadence Science
L
2 in.
gauge
14
General description
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)