Merck
Rapid urease screening of Yersinia on CIN agar plates.
International journal of food microbiology (2003-06-05)
Friederike Hilbert, Sigrid Mayrhofer, Frans J M Smulders
ABSTRACT
Yersinia enterocolitica is an important foodborne pathogen, but isolation of virulent Yersinia from food sources is still time consuming and requires skills. In this article, we describe a rapid urease screening on cefsulodin-irgasan-novobiocin (CIN) agar plates with an agar overlay assay. This test is simple to perform, all colonies on a plate can be checked simultaneously, it only takes minutes for detection of urease-positive colonies and the colonies survive for transfer, further characterisation, and storage. Additionally, this method is useful to isolate virulent (urease-positive and pYV harbouring) Y. enterocolitica from foodstuffs.
Sigma-Aldrich
Cefsulodin sodium salt hydrate, third-generation cephalosporin antibiotic
Pricing and availability is not currently available.