Merck
  • Molecular principles of assembly, activation, and inhibition in epithelial sodium channel.
Molecular principles of assembly, activation, and inhibition in epithelial sodium channel.
eLife (2020-07-31)
Sigrid Noreng, Richard Posert, Arpita Bharadwaj, Alexandra Houser, Isabelle Baconguis
ABSTRACT
The molecular bases of heteromeric assembly and link between Na+ self-inhibition and protease-sensitivity in epithelial sodium channels (ENaCs) are not fully understood. Previously, we demonstrated that ENaC subunits - α, β, and γ - assemble in a counterclockwise configuration when viewed from outside the cell with the protease-sensitive GRIP domains in the periphery (Noreng et al., 2018). Here we describe the structure of ENaC resolved by cryo-electron microscopy at 3 Å. We find that a combination of precise domain arrangement and complementary hydrogen bonding network defines the subunit arrangement. Furthermore, we determined that the α subunit has a primary functional module consisting of the finger and GRIP domains. The module is bifurcated by the α2 helix dividing two distinct regulatory sites: Na+ and the inhibitory peptide. Removal of the inhibitory peptide perturbs the Na+ site via the α2 helix highlighting the critical role of the α2 helix in regulating ENaC function.
Sigma-Aldrich
Amiloride hydrochloride hydrate, ≥98% (HPLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Benzamil hydrochloride hydrate, ≥98% (HPLC)