Skip to Content
Merck
All Photos(1)

Key Documents

1A01340

USP

2,3-Dimethyl-2,3-Butanediamine Dihydrochloride

Pharmaceutical Analytical Impurity (PAI)

Synonym(s):

2,3-Dimethyl-2,3-butanediamine dihydrochloride, 1,1,2,2-Tetramethylethylenediamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H16N2 · 2HCl
CAS Number:
Molecular Weight:
189.13
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.24

grade

pharmaceutical analytical impurity (PAI)

Agency

USP

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].[N+H3]C(C([N+H3])(C)C)(C)C

InChI key

BRLVXFONMZRJCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3-Dimethyl-2,3-Butanediamine Dihydrochloride is a USP Pharmaceutical Analytical Impurity (PAI).

USP PAI are a product line of impurities suitable for research and analytical purposes, which help to ensure the quality and safety of medicines.

For more information about this PAI, visit here.

Application

2,3-Dimethyl-2,3-Butanediamine Dihydrochloride is intended for use in analytical testing to detect, identify, and measure pharmaceutical impurities.

Features and Benefits

USP PAI advance your early analytical R&D and process development.

PAI can be used in the following applications:

1. Conduct analytical tests during early formulation feasibility studies.

2. Determine degradation impurities produced during stress studies.

3. Develop, validate, and transfer analytical methods.

4. Perform spiking studies during process R&D to demonstrate depletion upon recrystallization.

5. Record retention times and/or spectra.

6. Determine relative response factors.

7. Identify unknown impurities that formed during ICH stability conditions.

8. Identify impurities that are present in the Reference Listed Drug.

9. Test for and profile impurities not listed in drug substance and drug product monographs.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service