Skip to Content
Merck
All Photos(1)

Key Documents

S6699

Sigma-Aldrich

β-Synuclein P123H histidine-tagged human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE), lyophilized powder

Synonym(s):

SNCB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in E. coli

Quality Level

Assay

≥90% (SDS-PAGE)

form

lyophilized powder

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SNCB(6620)

Biochem/physiol Actions

Synucleins (α, β, γ-synoretin) are a family of small, highly conserved proteins expressed primarily in neurons and certain tumors. β-Synuclein is an acidic, neuronal protein considered to be a non-amyloidogenic homolog of α-synuclein. It has been postulated that β-synuclein could act as a physiological inhibitor of α-synuclein aggregation and that it might protect the central nervous system from the neurotoxic effects of α-synuclein. The mechanisms of β-synuclein neuroprotection may involve direct interaction between β-synuclein and Akt, and thus suggest that this signaling pathway could be a therapeutic target for neurological conditions associated with Parkinson and α-synuclein aggregation. Overexpression of β-synuclein mutants (P123H and V70M) in neuroblastoma cells results in enhanced lysosomal pathology suggesting a causative role for these missense mutations in neurodegeneration stimulation.

Physical form

Lyophilized from a 0.2 μm filtered solution.

Reconstitution

Reconstitute with 0.5 mL water (~1 mg/mL). Store in working aliquots at −20 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service