Skip to Content
Merck
All Photos(1)

Key Documents

BHK/AC9

85040103, hamster kidney, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

BHK/AC9, 85040103

biological source

hamster kidney

growth mode

Suspension

karyotype

2n = 44, pseudodiploid

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Hamster Syrian kidney

Cell Line Description

BHK/AC9 is derived from the parent line BHK-21 (Clone-13) (Sigma Catalogue number. 85011433). The cells have been adapted to grow in suspension and have the same virus susceptibility as the parent line.

Application

Virus replication studies

Culture Medium

GMEM + 2mM Glutamine + 5% Tryptose Phosphate Broth + 5-10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 2-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service