Key Documents
Z178063
Scienceware® heavy-duty test-tube rack
Holds 72 x 10-13 mm tubes
About This Item
UNSPSC Code:
41122804
NACRES:
NB.35
Pricing and availability is not currently available.
material
polypropylene
feature
Holds 72 x 10-13 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18855-0000
L × W × H
213 mm × 110 mm × 111 mm
General description
Natural PP, stackable. Steam autoclavable to 250°F (121°C). Rows lettered and numbered for easy identification. Base plate has rounded wells with drain holes; end plates with hand grips.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)