Merck
  • 2-Arylbenzo[b]arsoles: an experimental and computational study on the relationship between structural and photophysical properties.
2-Arylbenzo[b]arsoles: an experimental and computational study on the relationship between structural and photophysical properties.
Dalton transactions (Cambridge, England : 2003) (2020-09-24)
Kosuke Ishijima, Susumu Tanaka, Hiroaki Imoto, Kensuke Naka
ABSTRACT
Benzo[b]arsole derivatives, being arsenic analogues of indole, were synthesized by utilizing a safely prepared arsenic precursor. The structural and photophysical properties of the obtained 2-arylbenzo[b]arsoles were experimentally and computationally studied in comparison with those of 1,2,5-triarylarsoles and 9-phenylarsafluorene. It was found that the emission color and/or quantum yield were significantly affected by substituents on the luminescent center and metal-coordination to the arsenic atom. This is the first study on the structure-property relationship of benzo[b]arsole derivatives.
Sigma-Aldrich
4-Methoxyphenylboronic acid, ≥95.0%