SRP0585
Monkey CD46-BC, His-Tag
recombinant, expressed in HEK 293 cells
Synonym(s):
membrane co-factor protein
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.26
Recommended Products
recombinant
expressed in HEK 293 cells
form
frozen liquid
mol wt
34.2 kDa
GenBank accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
Monkey ... CD46(17221)
General description
Cynomolgus monkey (Macaca fascicularis) CD46-BC, also known as membrane co-factor protein, a.a. 35-328 with C-terminal His-tag, expressed in a HEK293 cell expression system. MW = 36.5 kDa (precursor), 34.2 kDa (mature). Note: this protein is expressed with a 34 amino acid signal sequence that is cleaved after secretion and is heavily glycosylated, resulting in a protein that runs at a higher apparent MW.
Physical form
Aqueous solution
Legal Information
GenBank is a registered trademark of United States Department of Health and Human Services
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service