Skip to Content
Merck
All Photos(1)

Documents

412400

Sigma-Aldrich

IQ-1

A cell-permeable tetrahydroisoquinolinylidene compound that modulates Wnt/β-catenin signaling by targeting the PR72/130 subunit of PP2A and thereby blocking PP2A/Nkd complex formation.

Synonym(s):

IQ-1, 2-(4-Acetylphenylazo)-2-(3,3-dimethyl-3,4-dihydro-2H-isoquinolin-1-ylidene)-acetamide, Wnt Pathway Activator III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22N4O2
CAS Number:
Molecular Weight:
362.42
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable tetrahydroisoquinolinylidene compound that modulates Wnt/β-catenin signaling by targeting the PR72/130 subunit of PP2A and thereby blocking PP2A/Nkd complex formation, resulting in diminished β-catenin/p300 interaction and a concomitant increase in β-catenin/CBP usage. Co-adminitration of IQ-1 (4 µg/ml) and Wnt3a (100 ng/ml), but neither reagent alone, has been shown to be sufficient in maintaining long-term (>48 days) murine ESCs (Embryonic Stem Cells) pluripotency in the absence of serum.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Miyabayashi, T., et al. 2007. Proc. Natl. Acad. Sci. USA104, 5668.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service