Key Documents
Z678775
FORTUNA® Polyfix® bottle top dispenser with amber glass cylinder
dispensing volume 10-50 mL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
manufacturer/tradename
Fortuna Graf 101.072-48
dispensing volume measuring range
10-50 mL
volume
10-50 mL
fitting
GL 40
GL 45
Related Categories
General description
For dispensing light sensitive liquids, weak acids and alkalis
Features:
  • With amber dispensing cylinder made of glass, coated with PP-protection against breakage
  • Glass piston
  • Ergonomic finger protection cap
  • Fast and easy volume setting with additional fine adjustment
  • FIX adapter made of PP
  • Valve block made of PP, no metal springs in the valve - no corrosion
Supplied as FORTUNA Polyfix dispenser, 2 PP adapters GL40 + GL45 & 1 PTFE intake tube
Legal Information
FORTUNA is a registered trademark of Poulten & Graf Ltd.
POLYFIX is a registered trademark of Poulten & Graf Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)