Skip to Content
Merck
All Photos(1)

Key Documents

WHA77009901

Whatman® UNIFILTER® 24 well filter plates

natural polypropylene, GF/C membrane, non-sterile, biomolecule, 25 ea

Synonym(s):

Z746584, micro plate, multiwell plate, well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900
NACRES:
NB.24

product name

Whatman® UNIFILTER® plates 24, GF/C, natural polypropylene, filter bottom with long drip director

material

glass microfiber membrane
polypropylene

sterility

non-sterile

feature

GF/C, natural polypropylene, filter bottom with long drip director

packaging

pkg of 25 ea

manufacturer/tradename

Whatman 7700-9901
Whatman Article No. 28420845 (US reference)

size

24 well

well maximum volume

1 μm

working volume

10 mL

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

The 10 mL UNIFILTER is used with large sample or reagent volumes. Typical applications include biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation.

Polypropylene microplate for biomolecule purification

Applications such as combinatorial chemistry library generation that require large sample volumes depend on microplates capable of filtering and holding these volumes. Biomolecule purification by solid phase extraction (SPE) is an example of one such an application.

Whatman UNIFILTER 24 well microplates are well suited for large sample volume filtration. As SPE plates, they possess high heat and chemical resistance, as well as novel long drip directors that minimize the risk of well-to-well cross-contamination.

Features and Benefits

  • Polypropylene microplate is heat and chemical resistant
  • 24 well microplate with 10 mL wells appropriate for large sample volumes
  • No cross talk: patented integral filter drip director design prevents well-to-well cross contamination
  • Filtration media options include glass microfiber, melt-blown polypropylene, PTFE, VFE

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

UNIFILTER is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service