707422
N-Fmoc-isocyanate
95%
Synonym(s):
N-(9-Fluorenylmethoxycarbonyl)isocyanate, Carbonisocyanatidic acid 9H-fluoren-9-ylmethyl ester, Fluoren-9-ylmethyloxycarbonylisocyanate
About This Item
Recommended Products
Assay
95%
form
solid
mp
52-56 °C
functional group
Fmoc
storage temp.
−20°C
SMILES string
O=C=NC(=O)OCC1c2ccccc2-c3ccccc13
InChI
1S/C16H11NO3/c18-10-17-16(19)20-9-15-13-7-3-1-5-11(13)12-6-2-4-8-14(12)15/h1-8,15H,9H2
InChI key
SRAHDGNWVCEFKK-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service