Skip to Content
Merck
All Photos(1)

Key Documents

272736

Sigma-Aldrich

Ethyl trans-α-cyanocinnamate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=C(CN)CO2C2H5
CAS Number:
Molecular Weight:
201.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder

bp

188 °C/15 mmHg (lit.)

mp

51-53 °C (lit.)

functional group

ester
nitrile
phenyl

SMILES string

CCOC(=O)\C(=C\c1ccccc1)C#N

InChI

1S/C12H11NO2/c1-2-15-12(14)11(9-13)8-10-6-4-3-5-7-10/h3-8H,2H2,1H3/b11-8+

InChI key

KCDAMWRCUXGACP-DHZHZOJOSA-N

Application

Ethyl trans-α-cyanocinnamate may be used in chemical synthesis studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service