Skip to Content
Merck
All Photos(1)

Key Documents

48246

Supelco

EPA 525, 525.1 PCB Mix

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Agency

EPA 525,525.1,525.2

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-30°C

Application

EPA 525, 525.1 PCB Mix has been used as a deuterated internal standard in the determination of polycyclic aromatic hydrocarbons in marine water samples using gas chromatography coupled with mass spectrometry (GC-MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    2-Chlorobiphenyl
    2,3-Dichlorobiphenyl
    2,2′,3,3′,4,4′,6-Heptachlorobiphenyl
    2,2′,4,4′,5,6′-Hexachlorobiphenyl
    2,2′,3′,4,6-Pentachlorobiphenyl
    2,2′,4,4′-Tetrachlorobiphenyl
    2,2′,3,3′,4,5′,6,6′-Octachlorobiphenyl
    2,4,5-Trichlorobiphenyl
    See All (8)

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dynamic behaviour of polycyclic aromatic hydrocarbons in Brighton marina, UK
King.J.A, et al.
Marine Pollution Bulletin, 48, 229-239 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service