Skip to Content
Merck
All Photos(10)

Key Documents

ZRB1920

Sigma-Aldrich

Anti-Nanog Antibody, clone 4D17 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Homeobox protein NANOG;ES cell-associated protein 4;Early embryo specific expression NK-type homeobox protein;Homeobox transcription factor Nanog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4D17, recombinant monoclonal

description

4D17 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 31.86 kDa
observed mol wt ~37 and 32 kDa

purified by

using Protein A

species reactivity

mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Nanog(71950)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 4D17 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Nanog. It targets an epitope within 20 amino acids from the N-terminal half. Predicted to react with both isoforms.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the N-terminal half of mouse Nanog.

Application

Quality Control Testing

Evaluated by Western Blotting in E14-TG2a cell lysates.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Nanog in E14-TG2a cell lysates.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Nanog in Mouse embryonic stem cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Nanog in mouse testis tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Nanog in E14-TG2a cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Homeobox protein NANOG (UniProt: Q80Z64; also known as ES cell-associated protein 4, Early embryo specific expression NK-type homeobox protein, Homeobox transcription factor Nanog) is encoded by the Nanog (also known as Ecat4, Enk) gene (Gene ID: 71950) in murine species. Nanog is a transcription factor that is involved in the self-renewal of embryonic stem cells (ES) and is a critical factor for the maintenance of the undifferentiated state of pluripotent cells. It binds optimally to the DNA consensus sequence 5′-TAAT[GT][GT]-3′ or 5′-[CG][GA][CG]C[GC]ATTAN[GC]-3′. Its expression is observed in inner cell mass (ICM) of the blastocyst and gonocytes and its expression decreases with ES differentiation. Nanog can autorepress its own expression in differentiating ES cells by binding to its own promoter following interaction with ZNF281/ZFP281, leading to recruitment of the NuRD complex and subsequent repression of expression. Overexpression of Nanog in ES cells maintains specific differentiation, however, its overexpression in cancer stem cells is reported to inhibit apoptosis. Together with OCT4, it can activate SNAIL2 and promote metastasis and the EMT phenotype in cancer cells. Nanog undergoes phosphorylation on serine 130 and 131 by casein kinase I and phosphomimetic substitution of serine 130 is shown to abolish its DNA binding and eliminates LIF‐independent self‐renewal. On the other hand, phosphomimetic substitution of serine 131 enhances LIF‐independent self‐renewal but does not affect its DNA binding. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Mullin, NP., et al. (2021). FEBS Lett. 595(1); 14-25; Gawlik-Rzemieniewska, N., and Bednarek, I (2016). Cancer Biol. Ther. 17(1); 1-10; Kim, S-H., et al. (2014). Stem Cell Res. 13(1); 1-11).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service