Skip to Content
Merck
All Photos(1)

Documents

Z742621

Tube-O-CONCENTRATOR Concentration Device

20 ‑ 250 μL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123100
NACRES:
NA.56

feature

20 ‑ 250 μL

packaging

pkg of 5 ea

manufacturer/tradename

G-BioSciences 786-625

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Tube-O-CONCENTRATOR is a versatile concentration device that utilizes a novel liquid polymer and our patented
Tube-O-CONCENTRATOR for the rapid concentration of dilute protein solutions with zero loss. The unique tube design ensures that 100% sample is recovered; simple place the entire device in a bench top centrifuge and spin for a few seconds.
The Tube-O-CONCENTRATOR solution is a liquid polymer that rapidly absorbs water through the dialysis membrane in the Tube-O-CONCENTRATOR cap, which retains all molecules with a molecular weight >1kDa.
Tube-O-CONCENTRATOR is available in two sizes for concentrating sample volumes of up to 250μl (Micro) or 2.5ml (Medi).
Include:

  • Concentrator Solution - 125 ml
  • Tube-O-DIALYZER, 1,000MWCO, Medi - sample volume up to 2.5 ml - 5 units
  • Micro Dialysis Cups - 5 units
  • Floats - 5 units
  • Storage Caps - 5 units

Legal Information

Tube-O-CONCENTRATOR is a trademark of Geno Technology, Inc.
Tube-O-DIALYZER is a trademark of Geno Technology, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

149.0 °F

Flash Point(C)

65 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service