Key Documents
Z689386
Jenco variable intensity halogen illuminator
output150 watts, AC/DC input 115V AC
About This Item
UNSPSC Code:
41100000
manufacturer/tradename
Jenco
output
150 watts
General description
The independent illuminators (Z689386) have a 150 or 30 Watt variable intensity halogen light source. Three different light guides are available to attach to this source: a flexible single gooseneck (Z689394), a bifurcated or dual gooseneck (Z689408) and a ring light (Z689416). The 30 Watt source will function only with a single gooseneck.
Choose from one of the most recent versions:
Certificates of Analysis (COA)