Z653802
Pyrex® glass gas distribution tubes
porosity grade 0
Synonym(s):
3830/00M
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass tube
colorless tube
sterility
non-sterile
feature
graduations: no
manufacturer/tradename
SciLabware 3830/00M
availability
available only in EU
Overall L
300 mm
thimble O.D.
15 mm
tube diam.
8 mm
8 mm
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
- Ideal for the dispersion of gas oin liquids
- Borosilicate glass tube with sintered thimble
- Choice of thimble porosities to achieve optimum gas bubble size
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service