Documents
TMO362002-0002
Nalgene® Boston round bottle without cap
natural high-density polyethylene, volume 60 mL, case of 1000 ea
Synonym(s):
HDPE bottle, nalgene bottle
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
natural high-density polyethylene
packaging
case of 1000 ea
manufacturer/tradename
Nalgene 362002-0002
capacity
60 mL
volume
60 mL
General description
Conduct demanding packaging applications with Thermo Scientific Nalgene Boston Round HDPE Bottles without Closure, which are both durable and reliable products. These high density polyethylene bottles are rigid and translucent with excellent chemical resistance. Uniform walls make these bottles resistant to splitting and puncturing.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries