Skip to Content
Merck
All Photos(1)

Documents

SML1702

Sigma-Aldrich

SW203668 trifluoroacetate

≥98% (HPLC)

Synonym(s):

4-(Amino(phenyl)methyl)-N-(6-methoxybenzo[d]thiazol-2-yl)benzamide trifluoroacetate, 4-(Aminophenylmethyl)-N-(6-methoxy-2-benzothiazolyl)-benzamide trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19N3O2S · CF3CO2H
CAS Number:
Molecular Weight:
503.49
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(C(O)=O)F.NC(C1=CC=CC=C1)C(C=C2)=CC=C2C(NC3=NC4=CC=C(OC)C=C4S3)=O

InChI

1S/C22H19N3O2S.C2HF3O2/c1-27-17-11-12-18-19(13-17)28-22(24-18)25-21(26)16-9-7-15(8-10-16)20(23)14-5-3-2-4-6-14;3-2(4,5)1(6)7/h2-13,20H,23H2,1H3,(H,24,25,26);(H,6,7)

InChI key

SIUPHFAIFLEFIT-UHFFFAOYSA-N

Biochem/physiol Actions

SW203668 is activated in cell lines expressing cytochrome P450 (CYP) 4F11 to an irreversible inhibitor of stearoyl CoA desaturase. Stearoyl CoA desaturase is not essential in the liver where SW203668 activation takes place, but is necessary for many tumor cells, several of which also express high levels of CYP4F11. SW203668 inhibited growth of lung carcinoma cell line H2122 with IC50 values of 0.029 μM and 0.007 μM, respectively for the (+) and (-) enantiomers. SW203668 also substantially reduced growth rate in H2122-derived tumors in immunodeficient mice.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service