Skip to Content
Merck
All Photos(1)

Documents

SCP0144

Sigma-Aldrich

[Tyr0]-Fibrinopeptide A human

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C72H106N20O28
Molecular Weight:
1699.73
UNSPSC Code:
12352202
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥70%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Tyr-Ala-Asp-Ser-Gly-Glu-Gly-Asp-Phe-Leu-Ala-Glu-Gly-Gly-Gly-Val-Arg

Application

[Tyr0]-Fibrinopeptide A (YADSGEGDFLAEGGGVR) is used as an N-terminally modified (tyr) analogue of Fibrinopeptide A (ADSGEGDFLAEGGGVR).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J F Galey et al.
Journal of inorganic biochemistry, 44(2), 149-153 (1991-11-01)
Results are reported for a potentiometric and spectroscopic (visible, CD, and EPR) study of the complexes of fibrinopeptide A (Ala-Asp-Ser-Gly-Glu-Gly-Asp-Phe-Leu-Ala-Glu-Gly-Gly-Gly-Val-Arg) with H+, Cu2+ and Zn2+. They show that the peptide forms stable complexes with Cu2+, largely as a result of
Ming-Gang Cheng et al.
Nan fang yi ke da xue xue bao = Journal of Southern Medical University, 28(6), 1005-1007 (2008-06-28)
To establish a high-performance capillary electrophoresis (HPCE)-based method for detection of trace amount of urinary fibrinopeptide A and B (FPA and FPB, respectively) as the specific molecular markers of thrombus formation in vivo. The HPCE system consisted of a 25

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service