Skip to Content
Merck
All Photos(2)

Documents

SAB5702880

Sigma-Aldrich

Anti-MOV10 Antibody, clone 1M7M9, Rabbit Monoclonal

Synonym(s):

fSAP113, gb110

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

1M7M9, monoclonal

form

liquid

mol wt

114 kDa

species reactivity

human, rat, rat

concentration

1.5 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

AATVTSYLKLLLAPSSKKGKARLSPRSVGVISPYRKQVEKIRYCITKLDRELRGLDDIKDLKVGSVEEFQGQERSVILISTVRSSQSFVQLDLDFNLGFLK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MOV10(4343)

General description

5′ to 3′ RNA helicase contributing to UPF1 mRNA target degradation by translocation along 3′ UTRs. Required for microRNA (miRNA-mediated gene silencing by the RNA-induced silencing complex (RISC. Required for both miRNA-mediated translational repression and miRNA-mediated cleavage of complementary mRNAs by RISC. In cooperation with FMR1, regulates miRNA-mediated translational repression by AGO2. Restricts retrotransposition of long interspersed element-1 (LINE-1 in cooperation with TUT4 and TUT7 counteracting the RNA chaperonne activity of L1RE1. Facilitates LINE-1 uridylation by TUT4 and TUT7. Required for embryonic viability and for normal central nervous system development and function. Plays two critical roles in early brain development: suppresses retroelements in the nucleus by directly inhibiting cDNA synthesis, while regulates cytoskeletal mRNAs to influence neurite outgrowth in the cytosol (By similarity. May function as a messenger ribonucleoprotein (mRNP clearance factor. Exhibits antiviral activity against dengue virus (DENV.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 800-900 of human MOV10 (Q9HCE1).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service