Documents
R1525
Sigma-Aldrich
Texan reagent reservoir for multichannel pipettes
with lid, non-sterile, pack of 100 ea (10 pavks of 10ea)
Synonym(s):
multichannel pipette reservoir, reagent reservoir
About This Item
UNSPSC Code:
41121522
NACRES:
NB.01
material
V-bottom bottom
colorless
polypropylene
sterility
non-sterile
packaging
pack of 100 ea (10 pavks of 10ea)
manufacturer/tradename
Excel Scientific RSV-L175-N10
capacity
175 mL
General description
Texan reagent reservoirs are reusable -- an environmentally sound alternative to disposable reagent reservoirs. Their extra-large capacity means fewer refills and time saved. The V-shaped basin is compatible with both eight- and twelve-place multiple pipettors with minimal reagent waste. Lids are removable -- attached with ball and socket hinges -- and latch tightly for secure storage of left-over reagents. With lids closed, Texan reagent reservoirs are stackable. A perimeter ridge on the lid prevents sliding. Construction is clear, chemical-resistant, autoclavable polypropylene. Molded graduation marks provide volume indication at 25, 50, 75, 100, 125, and 150 mL.
Legal Information
Texan is a trademark of Excel Scientific, Inc.
Visit the Document Library
Kathrin I Mohr et al.
MicrobiologyOpen, 5(2), 268-278 (2015-12-17)
Myxobacteria harbor an enormous potential for new bioactive secondary metabolites and therefore the isolation of in particular new groups is of great interest. The diversity of myxobacteria present in two ecological habitats, namely sand from Kiritimati Island and German compost