The kit is designed to detect total polyamines and is not specific for putrescine, spermidine, or spermine individually. The enzyme that metabolizes polyamines is selective for diamines over monoamines, so it is unlikely that chitosan will interfere significantly, unless it is present at high concentrations. The name of the enzyme is considered proprietary information.
MAK349
Total Polyamine Assay Kit (Fluorometric)
sufficient for 100 fluorometric tests
Synonym(s):
Fluorescent Polyamine Quantification Kit
About This Item
Recommended Products
usage
sufficient for 100 fluorometric tests
detection method
fluorometric
relevant disease(s)
cancer; aging/geriatric diseases
storage temp.
−20°C
General description
Features and Benefits
Suitability
Principle
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Flash Point(F)
188.6 °F
Flash Point(C)
87 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How specific is MAK349 in detecting putrescine, spermidine, and spermine? Can the kit be used for a sample containing chitosan?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service