Skip to Content
Merck
All Photos(1)

Key Documents

MAK277

Sigma-Aldrich

GAPDH Activity Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

Glyceraldehyde-3-phosphate Dehydrogenase Activity Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

detection method

colorimetric

relevant disease(s)

cancer

storage temp.

−20°C

General description

GAPDH (Glyceraldehyde-3-Phosphate Dehydrogenase; EC 1.2.1.12) catalyzes the conversion of Glyceraldehyde-3-Phosphate (GAP) to 1, 3-Bisphosphate Glycerate (BPG) and plays a key role in glycolysis. The enzyme is involved in cellular processes such as apoptosis, membrane trafficking, iron metabolism and nuclear translocation. GAPDH (housekeeping gene) expression is stable and constitutive. Deregulation of GAPDH activity is associated with abnormal cell proliferation and carcinogenesis. Accurate quantitation of GAPDH activity is important for diagnosing diseases and studying normal cellular physiology.

Application

GAPDH Activity Assay Kit has been used to measure the enzyme activity of glyceraldehyde-3-phosphate dehydrogenase in testis.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for use with various tissue and cell culture samples.

Principle

The GAPDH Activity Assay Kit provides a simple and sensitive method for monitoring GAPDH activity in various samples. GAPDH activity is determined in a coupled enzyme reaction in which GAP is converted to BPG by GAPDH. This results in a colorimetric (450 nm) product proportional to the enzymatic activity present. The assay is sensitive to 100 mUnits/mL. One unit of GAPDH activity is the amount of enzyme that will generate 1.0 mmole of NADH per minute at pH 7.2 at 37 °C.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samir El Qaidi et al.
Scientific reports, 11(1), 3834-3834 (2021-02-17)
Type III secretion system effector proteins have primarily been characterized for their interactions with host cell proteins and their ability to disrupt host signaling pathways. We are testing the hypothesis that some effectors are active within the bacterium, where they
Altered expression profile of glycolytic enzymes during testicular ischemia reperfusion injury is associated with the p53/TIGAR pathway: effect of fructose 1, 6-diphosphate.
May A L and Renno W M
PeerJ, 4, e2195-e2195 (2016)
May Al-Maghrebi et al.
PeerJ, 4, e2195-e2195 (2016-07-22)
Background. Testicular ischemia reperfusion injury (tIRI) is considered the mechanism underlying the pathology of testicular torsion and detorsion. Left untreated, tIRI can induce testis dysfunction, damage to spermatogenesis and possible infertility. In this study, we aimed to assess the activities
Catríona M Dowling et al.
Science advances, 7(3) (2021-02-02)
Triple-negative breast cancer (TNBC) is a subtype of breast cancer without a targeted form of therapy. Unfortunately, up to 70% of patients with TNBC develop resistance to treatment. A known contributor to chemoresistance is dysfunctional mitochondrial apoptosis signaling. We set

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service