Skip to Content
Merck
All Photos(1)

Documents

M8157

Supelco

TMD-8 hydrogen and hydroxide form

hydrogen and hydroxide form

Synonym(s):

Mixed Bed Resin hydrogen and hydroxide form

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

form

gel form

moisture

~55%

technique(s)

LPLC: suitable

matrix

cross-linked polystyrene (gel)

particle size

16-40 mesh (wet)

capacity

0.80 meq/g by dry weight

separation technique

mixed mode

Looking for similar products? Visit Product Comparison Guide

Components

1:1 mixture of strong cation and anion exchange resins

Quality

Resin is self-indicating: color changes from blue to amber when exchange capacity is reached.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnus Lundborg et al.
Biomacromolecules, 12(11), 3851-3855 (2011-10-01)
The structural analysis of polysaccharides requires that the sugar components and their absolute configurations are determined. We here show that this can be performed based on NMR spectroscopy by utilizing butanolysis with (+)- and (-)-2-butanol that gives the corresponding 2-butyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service