K0252
[D-Arg2]-Kyotorphin acetate salt
≥97% (TLC)
Synonym(s):
Tyr-D-Arg
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H22N5O4 · C2H4O2
CAS Number:
Molecular Weight:
396.42
MDL number:
UNSPSC Code:
12352200
Assay
≥97% (TLC)
storage temp.
−20°C
SMILES string
CC(O)=O.N[C@@H](Cc1ccc(O)cc1)C(=O)N[C@H](CCCNC(N)=N)C(O)=O
Biochem/physiol Actions
Analgesic effect is six times that of kyotorphin
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Studies on peptides. XCIV. Synthesis and activity of kyotorphin and its analogs.
H Yajima et al.
Chemical & pharmaceutical bulletin, 28(6), 1935-1938 (1980-06-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service