HUTR05134
MISSION® 3′UTR Lenti GoClone™
Powered by SwitchGear Genomics™, 3′UTR, human, KLK9
Synonym(s):
HUTR
About This Item
description
Functional Validation of miRNA Gene Targets
product line
MISSION®
concentration
≥1x105 VP/ml (via p24 assay)
human 3′UTR sequence
AGTCTACACCAGCGTATGCCACTACCTTGACTGGATCCAAGAAATCATGGAGAACTGAGCCCGCGCGCCACGGGGGCACCTTGGAAGACCAAGAGAGGCCGAAGGGCACGGGGTAGGGGGTTCTCGTAGGGTCCCAGCCTCAATGGTTCCCGCCCTGGACCTCCAGCTGCCCTGACTCCCCTCTGGACACTAAGACTCCGCCCCTGAGGCTCCGCCCCCTCACGAGGTCAAGCAAGACACAGTCGCGCCCCCTCG (3′UTR sequences are truncated after the first 255 bases)
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... KLK9(284366)
Application
Physical form
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service