Skip to Content
Merck
All Photos(1)

Documents

H9659

Sigma-Aldrich

Hepatitis C virus NS4 antigen−Fluorescein

~1 mg/mL, ≥95% (SDS-PAGE), recombinant, expressed in E. coli

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

recombinant

expressed in E. coli

Quality Level

Assay

≥95% (SDS-PAGE)

mol wt

19 kDa (plus 114 kDa β-Gal tag)

concentration

~1 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

hepatitis C virus ... HCVgp1(951475)

Biochem/physiol Actions

Positive control for HCV antibodies.

Other Notes

HCV NS4a+b [1658-1863]-Galactosidase-tagged, fluorescein conjugate.

Physical form

Solution in 8 M urea, 20 mM Tris-HCl, pH 8.0, and 10 mM β-mercaptoethanol

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service