Skip to Content
Merck
All Photos(1)

Documents

H2915

Sigma-Aldrich

HCK (230-497), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

JTK9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

156-212 nmol/min·mg

mol wt

~57 kDa

UniProt accession no.

relevant disease(s)

cancer (leukemia)

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HCK(3055)

General description

HCK (hematopoietic cell kinase), a protein-tyrosine kinase belonging to the Src family, is expressed in certain hemopoietic cells. It is especially prominent in cells of myeloid lineage, particularly mature granulocytes and monocytes.

Biochem/physiol Actions

HCK (hematopoietic cell kinase) gene is mapped to human chromosome 20q11-q12. This region on the chromosome urdergoes interstitial deletions in some acute myeloid leukemias and myeloproliferative disorders, thereby suggesting HCK participation in the pathogenesis of these conditions.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S F Ziegler et al.
Molecular and cellular biology, 7(6), 2276-2285 (1987-06-01)
Protein-tyrosine kinases are implicated in the control of cell growth by virtue of their frequent appearance as products of retroviral oncogenes and as components of growth factor receptors. Here we report the characterization of a novel human protein-tyrosine kinase gene
N Quintrell et al.
Molecular and cellular biology, 7(6), 2267-2275 (1987-06-01)
We have isolated cDNAs representing a previously unrecognized human gene that apparently encodes a protein-tyrosine kinase. We have designated the gene as HCK (hemopoietic cell kinase) because its expression is prominent in the lymphoid and myeloid lineages of hemopoiesis. Expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service