H1904
Herpes simplex virus I glycoprotein B Fragment 497-507
≥90% (HPLC)
Synonym(s):
Thr-Ser-Ser-Ile-Glu-Phe-Ala-Arg-Leu-Gln-Phe
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C59H91N15O18
CAS Number:
Molecular Weight:
1298.44
MDL number:
UNSPSC Code:
12352200
Assay
≥90% (HPLC)
storage temp.
−20°C
Biochem/physiol Actions
Activates CD8+ cytotoxic T lymphocytes.
Other Notes
An H-2kb T-cell epitope of HSV-I gB.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J P Vasilakos et al.
Journal of immunology (Baltimore, Md. : 1950), 150(6), 2346-2355 (1993-03-15)
Cytotoxic T cells were induced in vivo by immunizing C57BL/6 mice with a single dose of a short synthetic peptide representing amino acid residues 497-507 from HSV-1 glycoprotein B. Primed lymph node cells did not require in vitro boosting with
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service