GE80-1129-46
IEF sample application piece, for MultiPhor™ II
pack of 200 ea
All Photos(1)
About This Item
Recommended Products
packaging
pack of 200 ea
manufacturer/tradename
Cytiva 80112946
Related Categories
General description
Samples can be applied on the surface of the gels either by pipetting on pieces from cellulose-cotton, which are placed on the gel surface at the optimal pH location, or on to a silicone applicator strip with sample holes, laid on the gel surface.
Legal Information
MultiPhor is a trademark of Cytiva
Storage Class Code